Skip to Content
Merck
All Photos(1)

Key Documents

CLS480135

Corning® Microcentrifuge Individual Adapters

Synonym(s):

lab equipment accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.11
Pricing and availability is not currently available.

feature

Holds 0.2 mL tubes

packaging

pack of 6 ea

manufacturer/tradename

Corning 480135

technique(s)

centrifugation: suitable

compatibility

for use with 6765-HS
for use with 6766-HS
for use with 6767-HS
for use with Corning® LSE High Speed Microcentrifuges

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service