Skip to Content
Merck
All Photos(1)

Key Documents

CLS4155

Corning® DeckWorks low binding tips

1 - 300μL low binding pipet tips, graduated, reload System, natural, non-sterile, polypropylene, 96 tips/deck, 10 decks/pack, 4 packs/case

Synonym(s):

low binding pipet tips, low binding pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01
volume:
1—300 μL
sterility:
non-sterile
feature:
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Tip shape: round
packaging:
pack of 960
manufacturer/tradename:
Corning 4155
Pricing and availability is not currently available.

material

natural tip
polypropylene

sterility

non-sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduated
Tip shape: round

packaging

pack of 960

manufacturer/tradename

Corning 4155

volume range

1-300 μL

volume

1—300 μL

suitability

suitable for (quantitative assays in which sample binding can significantly alter results; specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.)

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® DeckWorks Low Binding Tips

Corning DeckWorks low binding tips are available in 10 uL, 200 uL and 300 uL sizes. Proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results.

Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides. This system is designed to allow you to maximize storage space and help reduce the production and waste of plastic materials. Both the hinged racks and reload decks within this system are manufactured with recycled plastic.

The pipet tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes.

Medical grade resins and production excellence allow for full certification as:

  • Nonpyrogenic
  • Human DNA-free
  • RNase-/DNase-free

Legal Information

Corning is a registered trademark of Corning, Inc.
DeckWorks is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service