Skip to Content
Merck
All Photos(1)

Key Documents

CLS3895

Corning® NBS 1536 well plates, without lids

1536 well plate, polystyrene, NBS surface, non-sterile, 50/cs

Synonym(s):

1536 multiwell plate, 1536 well microplate, 1536 well microtiter plate, 1536 well plate, 1536 well tissue culture treated plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
black polystyrene plate
flat bottom wells clear
polystyrene
size:
1536 wells

material

black polystyrene plate
flat bottom wells clear
polystyrene

sterility

non-sterile

feature

lid: no
skirt
plate format: 1536 well standard

packaging

pack of 10
case of 50 ×

manufacturer/tradename

Corning 3895

size

1536 wells

well volume

12.5 μL

well working volume

8 μL

color

clear

suitability

suitable for (fluorescent homogeneous assays)

binding type

NBS (non-binding surface)

General description

Corning® NBS 1536 Well Plates, Without Lids

  • Designed for fluorescent homogeneous assays
  • Compatible with standard readers and automation
  • Compatible for bar coding
  • Superior performance compared to competitor plates: Lower CVs, higher signal-to-noise ratios, and lower background fluorescence
  • Non-Binding Surface (NBS) can reduce nonspecific binding and improve signal to noise ratio.
  • Recommended working volume of up to 10μL

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service