Skip to Content
Merck
All Photos(1)

Key Documents

CLS3575BC

Corning® NBS 384 well microplates

NBS (non-binding surface), black polystyrene, flat bottom, non-sterile, with generic barcode, lid: no, pack of 10

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
black polystyrene
flat bottom
flat bottom black polystyrene wells
polystyrene
square black wells
size:
384 wells
Pricing and availability is not currently available.

material

black polystyrene
flat bottom
flat bottom black polystyrene wells
polystyrene
square black wells

sterility

non-sterile
non-sterile

feature

lid: no
lid: no
skirt
with generic barcode

packaging

pack of 10
case of 50 ea

manufacturer/tradename

Corning 3575BC

maximum volume

112 μL

size

384 wells

wells

384 wells

working volume

80 μL

color

black

suitability

suitable for (cell cuture)

binding type

NBS (non-binding surface)
low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Non-Binding surface (NBS) multiwell plates have a modified polymer surface (PS) which results in a non-ionic hydrophilic surface (polyethylene oxide-like), thereby minimizing bio-interaction and enhancing the signal-to-noise ratio. Results have demonstrated NBS′s ability to significantly reduce protein and nucleic acid binding to polymers (<2 ng/cm2; over 90% reduction of protein binding compare to PS & PP surface), maintain enzyme activity, and inhibit adhesion of a number of cell lines. These novel surface technologies for assay plates are ideal for homogeneous assays such as Scintillation Proximity Assay (SPA). In addition to a 20+ fold increase in signal-to-noise ratio, selected assays have also displayed improved sensitivity and specificity. The recommended working volume of up to 20 μL to 80 μL for 384 well plates.

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service