Skip to Content
Merck
All Photos(1)

Key Documents

CLS357486

Corning® Falcon® Battery for Pipet Controller (Set of 3),

Synonym(s):

Corning pipet accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.13
Pricing and availability is not currently available.

packaging

pkg of 3 ea

General description

Falcon Battery for Pipet Controller (Set of 3)

Application

Liquid Handling

Features and Benefits

Pipet controller accessory

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service