Key Documents
CLS10044
Corning® HYPERStack® stacking tray, orange, ABS material
no adhesives, low particulate
About This Item
UNSPSC Code:
41100000
NACRES:
NA.75
material
(Acrylonitrile Butadiene Styrene, ABS)
manufacturer/tradename
Corning 10044
General description
Features and Benefits:
More cells – 2.5x more cells per volumetric footprint
Closed system – No open fluid manipulations
Scalable product – Multiple size offerings
Ergonomic design – Easier manipulation
Fixed media volume – 0.2 mL/cm2 fills vessel for optimal handling
Innovative assembly – No adhesives, low particulate
Lower disposal cost – Less volumetric waste per growth area
Legal Information
Corning is a registered trademark of Corning, Inc.
HYPERStack is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)