CHA00011B
EUR-2019-nCoV RdRp D for detection of SARS-CoV-2 RNA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105500
NACRES:
NA.51
Recommended Products
form
solution (in 10 mM Tris, 1 mM EDTA)
packaging
pkg of 1 vial
pH range
7.5-8.5
shipped in
ambient
storage temp.
−20°C
General description
EUR-2019-nCoV Modified RdRp (formerly Discrimatory) Individual Assay is for qPCR (5′ nuclease assay) detection of SARS-CoV-2.
The product is primers and probes, the sequences of which were designed and made publicly available by Charité Institute of Virology – Universitätsmedizin Berlin.
For additional details, please visit SigmaAldrich.com/ncov.
The product is primers and probes, the sequences of which were designed and made publicly available by Charité Institute of Virology – Universitätsmedizin Berlin.
For additional details, please visit SigmaAldrich.com/ncov.
Features and Benefits
The forward and reverse primers in each tube come with the following features:
The Dual-Labeled Probe in each tube comes with the following features:
- DNA bases
- No modifications
- Desalt purified
- 10 nmol each
The Dual-Labeled Probe in each tube comes with the following features:
- DNA bases
- 6-FAM™ reporter
- BXQ (Block All Quencher—SigmaAldrich.com/bxq)
- HPLC purified
- 5nmol
Components
The product includes 1 individual tube of a 20X primer and probe pool for quantification of a SARS-CoV-2 gene. Assay included is Modified RdRp.
The forward primer, reverse primer, and probe are at final concentrations of 10, 10, and 5 μM, respectively.
Modified RdRp Assay:
Forward Primer AAATGGTCATGTGTGGCGGT
Reverse Primer GTTAAAAACACTATTAGCATAAGCAGTTGT
Dual-Labeled Probe CAGGTGGAACCTCATCAGGAGATGC
The forward primer, reverse primer, and probe are at final concentrations of 10, 10, and 5 μM, respectively.
Modified RdRp Assay:
Forward Primer AAATGGTCATGTGTGGCGGT
Reverse Primer GTTAAAAACACTATTAGCATAAGCAGTTGT
Dual-Labeled Probe CAGGTGGAACCTCATCAGGAGATGC
Legal Information
FAM is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service