Skip to Content
Merck
All Photos(1)

Key Documents

LB199

98062321, human blood, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

LB199, 98062321

biological source

human blood

packaging

tube of 5 μg 98062321-DNA-5UG
pkg of vial of cells 98062321-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lymphocyte, trichothiodystrophy, EBV-transformed

Cell Line Description

LB199 was derived from mononuclear cells from a peripheral blood sample from a 16 year old male. The patient was diagnosed with trichothiodystrophy and was shown to have a severe deficiency in excision repair of UV-irradiation damage. The defect was able to complement all known Xeroderma pigmentosum complementation groups and was not intragenic. This cell line is therefore a representative of a new excision-repair complementation group. The cell line was established by subsequent EBV transformation. Fibroblasts from the same individual have been designated TTD1BR. EBV-transformed lymphoblastoid cells from the mother are available as LB198 (Sigma Catalogue number. 98062320).

Application

DNA repair deficiency studies

Culture Medium

RPMI 1640 + 4mM Glutamine + 200 μg/ml Sodium Pyruvate (NaPy) + 15% Heat Inactivated (HI-FBS).

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; dilute every 1 -2 days to 3 x 100,000 cells/ml; 5% CO2; 37°C. Cells form floating aggregates.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service