Skip to Content
Merck
All Photos(1)

Documents

OE21 Cell Line human

96062201, human esophagus, Epithelial

Synonym(s):

JROECL 21 Cells, OE 21 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

OE21 Cell Line human, 96062201, Caucasian oesophageal squamous cell carcinoma

biological source

human esophagus

growth mode

Adherent

karyotype

Aneuploid

morphology

Epithelial

products

Not specified

receptors

MHC class I

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Caucasian oesophageal squamous cell carcinoma

Cell Line Description

OE21, also known as JROECL21, was established in 1993 from a squamous carcinoma of mid oesophagus of a 74 year-old male patient. The tumour was identified as pathological stage IIA(UICC) and showed moderate differentiation. HLA-A,-B and -C (MHC class I) are expressed constitutively, expression of ICAM-1 can be induced by treatment with interferon-gamma. OE21 cells express epithelial cytokeratins and are tumourigenic in nude mice.

Application

Study of oesophageal cancer

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 12
D16S539: 13
D5S818: 12,13
D7S820: 11,12
THO1: 8
TPOX: 8,11
vWA: 16,17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:8 i.e. seeding at 1x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service