Skip to Content
Merck
All Photos(1)

Documents

Hepa-1c1c7

95090613, mouse liver, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Hepa-1c1c7, 95090613

biological source

mouse liver

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Aryl hydrocarbon

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse hepatoma

Cell Line Description

Hepa-1c1c7 was cloned from the cell line Hepa-1cl which was obtained from Hepa-1. Hepa-1 was derived from the BW7756 hepatoma that developed in a C57L mouse. Hepa-1c1c7 cells express the aryl hydrocarbon (Ah) receptor and were shown to be highly inducible for cytochrome P450IA1. A variety of mutants from this cell line have been established.

Application

Study of Ah receptors, P450IA1 regulation

Culture Medium

α MEM without nucleosides + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service