Skip to Content
Merck
All Photos(1)

Documents

DMS 79

95062824, human lung, Irregular

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

DMS 79, 95062824

biological source

human lung

growth mode

Aggregates in suspension

karyotype

Aneuploid

morphology

Irregular

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lung small cell carcinoma

Cell Line Description

DMS79 has been established from pleural fluid of a patient with small cell carcinoma of the lung. The patient was treated with cytoxan, vincristine, methotrexate and radiation therapy. The cultures show a tendency for the modal chromosome number to decline. The production of adrenocorticotropin (ACTH), bombesin, calcitonin, β-endorphin, 17-β-estradiol, lipotropin, ocytocin-neurophysin, parathyroid hormone and somatstatin-like imuno-reactivity has been reported. The cells have also been reported to have the following cell surface antigens: Leu-7, Class I HLA, Class II HLA, Mg23 defined by the AML-2-23 monoclonal antibody and the AML-1-99 defined antigen. The cells have a receptor for epidermal growth factor (EGF) and produce tumours in nude athymic mice. Cells grow as irregularly shaped aggregates.

Application

Human lung tumour model, carcinogenesis studies.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) (Heat Inactivated).

Subculture Routine

Maintain cultures between 3 ? 9 x 100,000 cells/ml; 5% CO2; 37°C. Only dilute 1:2 as a maximum. This cell line grows as clumps in suspension which can be easily dispersed with pipetting. Trypsinisation is not necessary. Routine splitting can therefore

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service