Skip to Content
Merck
All Photos(1)

Documents

#W7079 REM human cell line

IHW Number: 9223, 94022552

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human blood

ethnicity

Caucasian

gender

not specified

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Looking for similar products? Visit Product Comparison Guide

General description

Consanguineous: No
Homozygous: No

Cell Line Description

This cell line is an Epstein-Barr Virus (EBV) transformed lymphoblastoid line and is part of the Human Leukocyte Antigen (HLA) Typed Collection maintained by the European Collection of Cell Cultures (ECACC). HLA-Typing Data is available for this cell line.

DNA Profile

A: 0201 0206
Cw: 03 07
B: 6701 40011

Serological Profile

HLA A: 2
HLA C: 10
HLA B: 60 67
Bw4/bw6: 6

Culture Medium

RPMI 1640 (R0883) + 2mM Glutamine (G7513) + 20% Foetal Bovine Serum (FBS) (F2442). Once a growing culture is established the FBS concentration can be reduced to 10%.

Subculture Routine

Maintain cultures between 3x105 - 2x106 cells/ml, 5% CO2, 37 °C. Upon resuscitation remove cryoprotectant - centrifuge at 300 x g for 5 minutes and re-suspend the cell pellet in fresh culture medium.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has restrictions and can only be supplied to academic and not-for-profit organisations. Nucleic acids can be supplied to all researchers including those from commercial organisations.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service