Skip to Content
Merck
All Photos(1)

Documents

F10-150.58

93100509, Reactivity: Anti-LFA-1, sheep

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0

Reactivity

Anti-LFA-1, sheep

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse BALB/c spleen

Cell Line Description

The antibody recognises the sheep LFA-1 molecule expressed by virtually all leukocytes. There is species cross-reactivity with goat.

Immunogen

Sheep lymphocytes

Application

FACS

Culture Medium

IDMEM + 2mM glutamine + 1% Hypoxanthine, Aminopterin, Thymidine (HAT) + 10% Foetal Bovine Serum (FBS) (may benefit from IL-6 in medium)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service