Skip to Content
Merck
All Photos(1)

Key Documents

BE10-7

93040125, rat brain, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

BE10-7, 93040125

biological source

rat brain

packaging

tube of 5 μg 93040125-DNA-5UG
pkg of vial of cells 93040125-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat, BDIX, brain, pre-malignant

Cell Line Description

Clone of the line BE10-Intermediate (Sigma Catalogue number. 93040119). The cells have elevated levels of tissue plasminogen activity but do not grow in agar or in BDIX rats. The cells should not be used beyond passage 24 as they start to form tumours.

Application

Study of carcinogenesis

Culture Medium

DMEM + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 to 1:20 i.e. seeding at 1-2x100,000 cells/25cm2 using 0.25% trypsin/EDTA; 10-15% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service