Skip to Content
Merck
All Photos(1)

Documents

F10-89-4

89062101, Reactivity: Anti-CD45 determinant 1, human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

NS1/1

Reactivity

Anti-CD45 determinant 1, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse NS1/1 x Mouse BALB/c spleen

Cell Line Description

The antibody recognises the human leukocyte common antigen which is found on cells from spleen, lymph nodes, 83% bone marrow cells and granulocytes. The antigen recognised by F10-89-4 is a glycoprotein with a molecular weight of 190kDa to 215kDa.

Immunogen

Human T lymphocytes

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 2-5x100,000 cells/ml; 5% CO2; 37°C. These cells require mouse macrophage feeder cells upon resuscitation

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service