Skip to Content
Merck
All Photos(1)

Documents

C7

Reactivity: Anti-LDL Receptors, human & bovine, 88090802

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0-Ag14

Reactivity

Anti-LDL Receptors, human & bovine

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0-Ag 14 x Mouse BALB/c spleen

Cell Line Description

The antibody binds to low density lipoprotein (LDL) receptors from bovine and human tissue. It does not react with receptors from the mouse, rat, Chinese hamster, rabbit or dog.

Immunogen

Partially purified LDL receptor from bovine adrenal cortex

Application

Study of hyper choloesterolaemia

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service