Recommended Products
Product Name
CHO-K1.Cl-0204, from hamster ovary, 15042903
biological source
hamster ovary
description
CHO-K1, FcγRIIa, Fc gamma receptor 2A, 131R, allotype
growth mode
Adherent
morphology
Polygonal
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Stably transfected with a pcDNA3.1/Hygro(+) based vector with a human FcγRIIa 131R cDNA insert that is expressed as a transmembrane protein with its native intracellular domain
Cell Line Description
Clone of CHO-K1 cells expressing human FcγRIIa of the 131R allotype asa transmembrane protein with its native intracellular domain
Culture Medium
MEM (M2279) + 2mM L-Glutamine (G7513) + 1% Non Essential Amino Acids(NEAA) (M7145) + 10% FBS (F2442) with 600ug/ml hygromycin B
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6. Seeding at 2-5 x 10,000 cells/cm² using 0.05% trypsin/EDTA; culture conditions 5% CO2/95%air; 37°C
Other Notes
Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
If use of this culture results in a scientific publication, it should be cited in the publication as: CHO-K1.CI-0204 (ECACC 15042903)
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service