N2102Ep Clone 2/A6
6011803, human embryo, Not specified
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
Product Name
N2102Ep Clone 2/A6, 06011803
biological source
human embryo
growth mode
Adherent
karyotype
Not specified
morphology
Not specified
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human embryonal carcinoma
Cell Line Description
A non seminomatous germ cell carcinoma derived cell line. This cell line was established from a single cell clone from a subline of cell line 2102Ep passaged through a nude mouse. This cell line is tumourigenic in nude mice.
Culture Medium
DMEM (High Glucose) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS). No sodium pyruvate.
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:5 i.e. seeding at >6x10,000 cells/cm2 using 0.25% trypsin or trypsin or trypsin/EDTA; 10% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service