Skip to Content
Merck
All Photos(1)

Key Documents

C6607

Sigma-Aldrich

Caspase 10 human

>90% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, 8,000 units/mg protein (Bradford)

Synonym(s):

Flice2, Mch4

Sign Into View Organizational & Contract Pricing

Select a Size

25 UNITS
€308.00
100 UNITS
€629.00

€308.00


Estimated to ship on19 April 2025


Request a Bulk Order

Select a Size

Change View
25 UNITS
€308.00
100 UNITS
€629.00

About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

€308.00


Estimated to ship on19 April 2025


Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

Assay

>90% (SDS-PAGE)

form

lyophilized powder

specific activity

8,000 units/mg protein (Bradford)

mol wt

~30 kDa

solubility

PBS: soluble

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CASP10(843)

Biochem/physiol Actions

Caspase 10 has two death effector domains (DEDs) that bind to the DED in the adapter molecule FADD and recruits both TNFR1 and CD95 to form complexes with these receptors. Caspase 10 cleaves and activates caspases 3, 4, 6, 7, 8 and 9 which causes the proteolytic cleavage of many key proteins such as PARP. Based on gene expression studies, caspase 10 may be crucial in embryonic development. In view of its structural homology to caspase 8, the initiator caspase in death receptor-mediated apoptosis, caspase 10 may function in a similar and redundant manner.

Unit Definition

One unit will hydrolyze 1 nmol of the caspase substrate IETD-pNA to IETD and p-nitroaniline per hour at pH 7.2 at 37 °C.

Physical form

Lyophilized powder containing 0.052% ammonium sulfate, 0.158% Tris−HCl, and 0.76% sodium chloride.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caspases: the executioners of apoptosis.
Cohen, G.M.
Biochemistry, 326 (part 1), 1-1 (1997)
Marcin Poreba et al.
Nature protocols, 12(10), 2189-2214 (2017-09-22)
Many biologically and chemically based approaches have been developed to design highly active and selective protease substrates and probes. It is, however, difficult to find substrate sequences that are truly selective for any given protease, as different proteases can demonstrate
Marcin Poreba et al.
Nature protocols, 12(10), 2189-2214 (2017-09-22)
Many biologically and chemically based approaches have been developed to design highly active and selective protease substrates and probes. It is, however, difficult to find substrate sequences that are truly selective for any given protease, as different proteases can demonstrate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service