Skip to Content
Merck
All Photos(1)

Key Documents

C6111

Sigma-Aldrich

Anti-m-Calpain (Domain III) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

immunohistochemistry (frozen sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot (chemiluminescent): 1:5,000
western blot: 1:1,000

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CAPN3(825)
mouse ... Capn3(12335)
rat ... Capn3(29155)

Specificity

Recognizes latent and active m-calpain. Binds the reduced and non-reduced protein. Identifies bands at 80 kDa, 58 kDa, and a series of further cleaved active forms.

Immunogen

synthetic peptide corresponding to the C-terminal end of domain-III of the large subunit of human m-calpain (calpain-II, protein kinase C activating factor, E.C. 3.4.22.17).

Physical form

Solution in 0.01 M phosphate buffered saline, containing 50% glycerol and 0.05% sodium azide.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service