Skip to Content
Merck
All Photos(1)

Key Documents

C5863

Sigma-Aldrich

Anti-Calcium Channel (β4 Subunit) (Voltage Gated Ca2+ Channel) antibody produced in rabbit

~1 mg/mL, fractionated antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

western blot: 5-10 μg/mL using brain tissue lysate

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CACNB4(785)
mouse ... Cacnb4(12298)
rat ... Cacnb4(58942)

General description

The gene CACNB4 (Calcium channel voltage-dependent subunit beta 4) is mapped to human chromosome 2q22-23. It is present at the membrane as well as the nucleus.

Specificity

Specifically recognizes a splice variant of β4 (55 kDa).

Immunogen

synthetic peptide derived from the rat β4 calcium channel subunit conjugated to KLH.

Application

Anti-Calcium Channel (β-4 subunit) antibody produced in rabbit is suitable for western blot at a concentration of 5-10μg/mL using brain tissue lysate.
The antibody has been used for:
  • western blotting of proteins extracted from different brain regions of the mouse
  • western blotting of proteins isolated from primary culture of mouse cerebral cortical neurons

Biochem/physiol Actions

The CACNB4 (Calcium channel voltage-dependent subunit β 4) is an intracellular regulatory subunit of voltage-activated calcium channels that has significant impact on membrane expression and electrophysical characteristics of the channel. Calcium channel β subunits regulate voltage-dependent calcium currents through direct interaction with α1 subunits. Mutations in CACNB4 are associated with idiopathic generalized epilepsy and episodic ataxia. CACNB4 interacts with heterochromatin protein 1γ (HP1γ). The interaction attenuates the gene silencing activity by HP1.

Physical form

Solution in phosphate buffered saline containing 0.08% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masashi Katsura et al.
Journal of pharmacological sciences, 102(2), 221-230 (2006-10-13)
Previous reports revealed up-regulation of L-type high voltage-gated calcium channels (HVCCs) in mouse brains with ethanol physical dependence. We investigated mechanisms of enhancement of L-type HVCC function using mouse cerebrocortical neurons exposed to 50 mM ethanol for 3 days and
Masahiro Shibasaki et al.
Journal of pharmacological sciences, 105(2), 177-183 (2007-10-06)
As functional changes in L-type high voltage-gated calcium channels (HVCCs) are recognized to be one of the major neurochemical modifications occurring in brains of animals with morphine physical dependence, this study attempts to examine whether regional difference in the expressions
Solmaz Etemad et al.
Channels (Austin, Tex.), 8(4), 334-343 (2014-05-31)
Voltage-gated calcium channels regulate gene expression by controlling calcium entry through the plasma membrane and by direct interactions of channel fragments and auxiliary β subunits with promoters and the epigenetic machinery in the nucleus. Mutations of the calcium channel β(4)
A Escayg et al.
American journal of human genetics, 66(5), 1531-1539 (2000-04-14)
Inactivation of the beta4 subunit of the calcium channel in the mouse neurological mutant lethargic results in a complex neurological disorder that includes absence epilepsy and ataxia. To determine the role of the calcium-channel beta4-subunit gene CACNB4 on chromosome 2q22-23
Xingfu Xu et al.
The Journal of biological chemistry, 286(11), 9677-9687 (2011-01-12)
The β subunits of voltage-gated Ca(2+) channels are best known for their roles in regulating surface expression and gating of voltage-gated Ca(2+) channel α(1) subunits. Recent evidence, however, indicates that these proteins have a variety of Ca(2+) channel-independent functions. For

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service