Skip to Content
Merck
All Photos(1)

Key Documents

C1368

Sigma-Aldrich

Cyclopropanecarboxylic acid {3-[4-(3-trifluoromethyl-phenylamino)-pyrimidin-2-ylamino]-phenyl}-amide

≥98% (HPLC), solid

Synonym(s):

Aurora Kinase Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18F3N5O
CAS Number:
Molecular Weight:
413.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

H2O: <2 mg/mL
DMSO: >7 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cccc(Nc2ccnc(Nc3cccc(NC(=O)C4CC4)c3)n2)c1

InChI

1S/C21H18F3N5O/c22-21(23,24)14-3-1-4-15(11-14)26-18-9-10-25-20(29-18)28-17-6-2-5-16(12-17)27-19(30)13-7-8-13/h1-6,9-13H,7-8H2,(H,27,30)(H2,25,26,28,29)

InChI key

RDTDWGQDFJPTPD-UHFFFAOYSA-N

Biochem/physiol Actions

ATP-competitive Aurora A kinase inhibitor with IC50 = 42 nM

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pooja Mohan et al.
Oncotarget, 4(1), 80-93 (2013-01-19)
Malignant peripheral nerve sheath tumours (MPNST) are rare, hereditary cancers associated with neurofibromatosis type I. MPNSTs lack effective treatment options as they often resist chemotherapies and have high rates of disease recurrence. Aurora kinase A (AURKA) is an emerging target
Igor Astsaturov et al.
Science signaling, 3(140), ra67-ra67 (2010-09-23)
Intrinsic and acquired cellular resistance factors limit the efficacy of most targeted cancer therapeutics. Synthetic lethal screens in lower eukaryotes suggest that networks of genes closely linked to therapeutic targets would be enriched for determinants of drug resistance. We developed
Mahendra K Singh et al.
Cancer research, 70(21), 8907-8916 (2010-10-14)
Elevated expression of the NEDD9/HEF1/Cas-L scaffolding protein promotes tumor cell invasion and metastasis in multiple cancer cell types. Conversely, generation of mammary tumors in the mouse mammary tumor virus (MMTV)-polyoma virus middle T (PyVT) genetic model is delayed by a
Qiong Zhang et al.
Journal of the American Chemical Society, 128(7), 2182-2183 (2006-02-16)
The epidermal growth factor receptor (EGFR) tyrosine kinase was one of the first receptor tyrosine kinases to be targeted for drug development by the pharmaceutical industry due to its ubiquitous overexpression in a variety of tumors. Despite the validation of
Ming Shen Tham et al.
Nature communications, 15(1), 371-371 (2024-01-09)
Aurora Kinase A (AURKA) promotes cell proliferation and is overexpressed in different types of polycystic kidney disease (PKD). To understand AURKA's role in regulating renal cyst development we conditionally deleted the gene in mouse models of Autosomal Dominant PKD (ADPKD)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service