Skip to Content
Merck
All Photos(1)

Key Documents

C1307

Sigma-Aldrich

Cyclo(D-Glu-Ala-D-allo-Ile-Leu-D-Trp)

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H44N6O7
CAS Number:
Molecular Weight:
612.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

form

solid

composition

Peptide content, >60%

storage temp.

−20°C

SMILES string

CCC(C)C1NC(=O)C(C)NC(=O)C(CCC(O)=O)NC(=O)C(Cc2c[nH]c3ccccc23)NC(=O)C(CC(C)C)NC1=O

InChI

1S/C31H44N6O7/c1-6-17(4)26-31(44)36-23(13-16(2)3)29(42)35-24(14-19-15-32-21-10-8-7-9-20(19)21)30(43)34-22(11-12-25(38)39)28(41)33-18(5)27(40)37-26/h7-10,15-18,22-24,26,32H,6,11-14H2,1-5H3,(H,33,41)(H,34,43)(H,35,42)(H,36,44)(H,37,40)(H,38,39)

InChI key

UMKHUVRCCMAOBN-UHFFFAOYSA-N

Biochem/physiol Actions

Selective ETA endothelin receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Hiramatsu et al.
Circulation, 92(9 Suppl), II400-II404 (1995-11-01)
Prior studies suggest an important role for coronary endothelium in ischemia/reperfusion (I/R) injury. Decreased endothelial release of the vasodilator nitric oxide occurs after I/R, but the role of the endothelium-derived vasoconstrictor endothelin-1 (ET-1) in I/R is unknown. We measured plasma
N Syed et al.
The Journal of urology, 159(2), 563-566 (1998-07-02)
Endothelin-1 (ET-1), a peptide produced by the vascular endothelium, causes profound renal vasoconstriction by binding to ET-A receptors. The present study examined the renal actions of ET-1 after ET-A receptors were blocked by BE-18257B to unmask the functions of ET-B
[Agonists and antagonists to endothelin receptors].
M Ihara et al.
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme, 38(11), 1827-1840 (1993-08-01)
T Satoh et al.
FEBS letters, 310(1), 83-87 (1992-09-21)
Computer-aided molecular modelling of the endothelin (ET-A) receptor antagonists, BQ-123 and BE-18257B, shows that they have very similar 3D structures. Parts of their 3D structures are also shown to match closely with that reported for residues 6-8 in endothelin-1. On
K Kojiri et al.
The Journal of antibiotics, 44(12), 1342-1347 (1991-12-01)
Two endothelin (ET)-binding inhibitors, BE-18257A and BE-18257B, which antagonized 125I-ET-1 binding to a porcine aortic smooth muscle membrane, were isolated from the mycelium of a strain of Streptomyces misakiensis. These binding inhibitors were extracted with methanol from mycelium and purified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service