C111040
DMT-dC(bz) Phosphoramidite
configured for MerMade
Synonym(s):
Cytidine, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxy-, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite], DMT-dC(bz) amidite, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxycytidine, 3′-[2-cyanoethyl N,N-?bis(1-methylethyl)phosphoramidite], N4-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]
About This Item
Recommended Products
biological source
non-animal source (no BSE/TSE risk)
Quality Level
type
for DNA synthesis
product line
Proligo Reagents
Assay
≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)
form
powder or granules
technique(s)
oligo synthesis: suitable
impurities
≤0.3 wt. % water content (Karl Fischer)
color
white to off-white
λ
conforms (UV/VIS Identity)
compatibility
configured for MerMade
nucleoside profile
base: deoxycytidine
base protecting group: benzoyl
2' protecting group: none
5' protecting group: DMT
deprotection: standard
storage temp.
2-8°C
SMILES string
COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=CC(NC(=O)c4ccccc4)=NC3=O)(c5ccccc5)c6ccc(OC)cc6
InChI
1S/C46H52N5O8P/c1-32(2)51(33(3)4)60(57-29-13-27-47)59-40-30-43(50-28-26-42(49-45(50)53)48-44(52)34-14-9-7-10-15-34)58-41(40)31-56-46(35-16-11-8-12-17-35,36-18-22-38(54-5)23-19-36)37-20-24-39(55-6)25-21-37/h7-12,14-26,28,32-33,40-41,43H,13,29-31H2,1-6H3,(H,48,49,52,53)/t40-,41+,43+,60?/m0/s1
InChI key
PGTNFMKLGRFZDX-SALLYJDFSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Recommended cleavage and deprotection conditions are 8 hours at 55 °Cor 24 hours at room temperature using concentrated ammonia solution,for standard base-protected oligonucleotides
- The high coupling efficiency of Proligo′s DNA phosphoramidites leads tohigh-yield and high-quality oligonucleotides
Packaging
C111040-10G packaged in a 8 oz bottle with a 28-400 cap
C111040-6x5G packaged in a 8 oz bottle with a 28-400 cap
C111040-6x10G packaged in a 8 oz bottle with a 28-400 cap
C111040-20x10G packaged in a 1 L bottle with GL 45
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service