Key Documents
BMSH3565180HD
Sigma-Aldrich
Benchmark SureTherm 180L CO2 Incubator
AC/DC input 120 V, with High Heat Function
Synonym(s):
carbon dioxide incubator
About This Item
UNSPSC Code:
41102404
Pricing and availability is not currently available.
material
stainless steel
feature
with High Heat Function
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific H3565-180HD
parameter
temp. range at +5-60 °C (ambient)
0-20% (CO2 range)
inside W × L × H
47.3 cm (18.7 in.) × 52.8 cm (20.8 in.) × 71 cm (28 in.)
General description
Cell cultures depend on optimized growth conditions in order to thrive. With precise temperature and CO2 control, Benchmark′s SureTherm Incubators provide the perfect environment for your cultures. All SureTherm Incubators feature a proprietary heat distribution system that combines six sided heating with a low speed internal fan. Air is circulated uniformly throughout the chamber without air exchange from the external environment. This results in a controlled, protected environment with minimal temperature fluctuations (within 0.1°C). The 180L model also includes options for high heat decontamination, a split glass door and oxygen gas control.
These are the first and only incubators with the optional (patent pending) IncuView LCI (Live Cell Imaging) technology for viewing cells in “real time”. An internal microscope and large, external display allow cells to be viewed without removal from the incubator, eliminating the inherent temperature fluctuations and risk of contamination. Cells are easily viewed on the large, integrated display or viewed locally or remotely with a wifi enabled phone, tablet or computer. Cell viewing requires the IncuView Integrated Microscope Shelf.
Features and Benefits
  • 6 sided heating
  • Live cell imaging option
  • proprietary heat distribution system
  • high heat decontamination
Choose from one of the most recent versions:
Certificates of Analysis (COA)