Skip to Content
Merck
All Photos(1)

Key Documents

BM0017

Sigma-Aldrich

BMS-199264 hydrochloride

≥98% (HPLC)

Synonym(s):

(3S,4R)-4-[(4-Chlorophenyl)(1H-imidazol-2-ylmethyl)amino]-3,4-dihydro-2,2-dimethyl-6-(1-piperidinylsulfonyl)-2H-1-Benzopyran-3-ol hydrochloride, (3S-trans)-1-[[4-[(4-Chlorophenyl)(1H-imidazol-2-ylmethyl)amino]-3,4-dihydro-3-hydroxy-2,2-dimethyl-2H-1-benzopyran-6-yl]sulfonyl]-piperidine monohydrochloride, BMS 199264 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H31ClN4O4S · HCl
CAS Number:
Molecular Weight:
567.53
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

O=S(C1=CC=C(OC(C)(C)[C@@H](O)[C@@H]2N(CC3=NC=CN3)C4=CC=C(Cl)C=C4)C2=C1)(N5CCCCC5)=O.[H]Cl

Biochem/physiol Actions

BMS-191264 decreases cardiac necrosis and improves the recovery of contractile activities after reperfusion.[1]
BMS-199624 is a potent inhibitor of the ATP hydrolase activity of mitochondrial F1F0 ATP synthase.
BMS-199624 is a potent inhibitor of the ATP hydrolase activity of mitochondrial F1F0 ATP synthase. The compound BMS-199624 has no affect on the ATP synthase function of F1F0. In isolated rat hearts, BMS-199624 blocks depletion of ATP levels, and blocks necrosis during ischemia.

Features and Benefits

BMS-199264 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gary J Grover et al.
Cardiovascular therapeutics, 26(4), 287-296 (2008-11-28)
The mitochondrial F1F0 ATP synthase is responsible for the majority of ATP production in mammals and does this through a rotary catalytic mechanism. Studies show that the F1F0 ATP synthase can switch to an ATP hydrolase, and this occurs under

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service