Skip to Content
Merck
All Photos(1)

Documents

B7888

Sigma-Aldrich

Borneol

~75%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

~75%

SMILES string

CC1(C)[C@H]2CC[C@]1(C)[C@H](O)C2

Gene Information

human ... UGT1A4(54657)

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meimei Duan et al.
Pharmaceutical biology, 54(12), 3009-3018 (2016-07-20)
Selectively opening the blood-tumour barrier (BTB) is critical to deliver antitumour drugs from blood to tumour tissues. The BTB problem is attributed to the tight junctions (TJs), which consist of several transmembrane proteins. To investigate whether borneol could open the
Jing-Ya Chen et al.
Chemico-biological interactions, 261, 96-102 (2016-11-28)
CYP2B subfamily accounts for 2-10% of total hepatic CYP450 enzymes and participate in the metabolism of around 8% of clinical drugs. Borneol has been widely used in traditional Chinese medicine for thousands of years. There are many studies about borneol-induced
Ge Ru et al.
Drug delivery, 23(9), 3417-3423 (2016-05-14)
Although nanocarriers provide promising potential for oral drug delivery, the delivery efficiency remains unsatisfactory and needs to be improved. Size is considered to be the most important characteristic of nanoparticles related to their oral absorption. Borneol has been proved to
Ting Li et al.
AMB Express, 6(1), 109-109 (2016-11-11)
Vulvovaginal candidiasis (VVC) is an opportunistic fungal infection predominantly caused by Candida albicans affecting a significant number of women of reproductive age. The Chinese medicine, the Baofukang suppository is widely used in the clinic for its antimicrobial activity and is
Justine E Giffen et al.
Phytochemical analysis : PCA, 28(1), 16-26 (2016-12-16)
The Salvia genus contains numerous economically important plants that have horticultural, culinary and nutraceutical uses. They are often similar in appearance, making species determination difficult. Species identification of dried Salvia products is also challenging since distinguishing plant morphological features are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service