Skip to Content
Merck
All Photos(1)

Key Documents

B1647

Sigma-Aldrich

[Lys3]-Bombesin

≥97% (HPLC)

Synonym(s):

pGlu-Gln-Lys-Leu-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C71H110N22O18S
CAS Number:
Molecular Weight:
1591.83
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

product name

[Lys3]-Bombesin, ≥97% (HPLC)

Assay

≥97% (HPLC)

form

powder

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(CC(C)C)NC(=O)C(Cc1cnc[nH]1)NC(=O)CNC(=O)C(NC(=O)C(C)NC(=O)C(Cc2c[nH]c3ccccc23)NC(=O)C(CCC(N)=O)NC(=O)C(CC(N)=O)NC(=O)CNC(=O)C(CC(C)C)NC(=O)C(CCCCN)NC(=O)C(CCC(N)=O)NC(=O)C4CCC(=O)N4)C(C)C)C(N)=O

InChI

1S/C71H110N22O18S/c1-35(2)25-48(90-63(103)44(15-11-12-23-72)87-65(105)46(16-19-53(73)94)88-64(104)45-18-21-56(97)83-45)62(102)79-32-57(98)85-52(29-55(75)96)70(110)89-47(17-20-54(74)95)66(106)92-50(27-39-30-78-42-14-10-9-13-41(39)42)67(107)82-38(7)61(101)93-59(37(5)6)71(111)80-33-58(99)84-51(28-40-31-77-34-81-40)69(109)91-49(26-36(3)4)68(108)86-43(60(76)100)22-24-112-8/h9-10,13-14,30-31,34-38,43-52,59,78H,11-12,15-29,32-33,72H2,1-8H3,(H2,73,94)(H2,74,95)(H2,75,96)(H2,76,100)(H,77,81)(H,79,102)(H,80,111)(H,82,107)(H,83,97)(H,84,99)(H,85,98)(H,86,108)(H,87,105)(H,88,104)(H,89,110)(H,90,103)(H,91,109)(H,92,106)(H,93,101)

InChI key

DXMFSGIDMUPBCC-UHFFFAOYSA-N

Gene Information

human ... GRP(2922)

Amino Acid Sequence

Glp-Gln-Lys-Leu-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2

Biochem/physiol Actions

[Lys3]-Bombesin and [Tyr4]-Bombesin are Bombesin analogues used to differentiate and study the bombesin receptors BBR1, BBR2 and BBR3 and the homologue receptors, gastrin-releasing peptide (GRP) receptor (GRPR).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Fleischmann et al.
Laboratory investigation; a journal of technical methods and pathology, 80(12), 1807-1817 (2001-01-05)
Overexpression of receptors for regulatory peptides in various human diseases is reportedly of clinical interest. Among these peptides, bombesin and gastrin-releasing peptide (GRP) have been shown to play a physiological and pathophysiological role in pancreatic tissues. Our aim has been
J B Trepel et al.
The Biochemical journal, 255(2), 403-410 (1988-10-15)
Bombesin is an amphibian tetradecapeptide whose mammalian homologue, gastrin-releasing peptide (GRP), is produced by many small-cell lung-cancer (SCLC) cells, and which can function in an autocrine growth-promoting manner in SCLC. Studies reported here show that [Tyr4]bombesin and its congeners increase
Edyta Podstawka
Biopolymers, 89(6), 506-521 (2007-12-22)
This work describes the molecular structure of bombesin (BN) and its analogs on the basis of the absorption infrared and Raman results described below. In these analogues is replaced one ([D-Phe12]BN, [Tyr4]BN, and [Lys3]BN) or two ([Tyr4,D-Phe12]BN, [D-Phe12,Leu14]BN, and [Leu13-(R)-Leu14]BN)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service