Skip to Content
Merck
All Photos(1)

Key Documents

B0311

Sigma-Aldrich

Bmx Active human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

Bone Marrow Kinase X-Linked

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

Assay

≥85% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

≥10 units/mg protein

mol wt

~110 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BMX(660)

Biochem/physiol Actions

The BMX gene encodes a novel non-receptor tyrosine kinase, which may play a role in the growth and differentiation of hematopoietic cells. Bmx induces the activation of the Stat-signaling pathway.

Unit Definition

Kinase activity is measured as the molar amount of phosphate incorporated into Poly Glu-Tyr substrate per minute per mg protein at 30 °C using a final concentration of 50 μM [32P]-ATP.

Physical form

Solution of 50 μL in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, and 30% glycerol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service