AXYTF400
Pipette filter tips
for Eppendorf® pipettors, ultramicro, size 0.5-10 μL, pkg of 10x1000tips/cs, sterile
Synonym(s):
pipet tips
About This Item
RNase free
barrier: no
endotoxin free
filter
Corning
Recommended Products
material
clear
sterility
sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter
manufacturer/tradename
Axygen Inc TF-400
Corning
packaging
pkg of 10x1000tips/cs
size
0.5-10 μL
volume
0.5—10 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service