Skip to Content
Merck
All Photos(1)

Key Documents

AXYRFLR

Empty Pipette tip refill rack, with lid

for use with P2/P10 Gilson-style and 200 μL universal fit tip inserts, pkg of 5x10racks/cs

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.02
capacity:
200 μL
packaging:
pkg of 5x10racks/cs
manufacturer/tradename:
Axygen Inc RFL-R
Corning
Pricing and availability is not currently available.

manufacturer/tradename

Axygen Inc RFL-R
Corning

packaging

pkg of 5x10racks/cs

capacity

200 μL

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service