Skip to Content
Merck
All Photos(2)

Documents

AV37665

Sigma-Aldrich

Anti-GPC3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Glypican 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

64 kDa

species reactivity

guinea pig, horse, canine, rat, mouse, rabbit, bovine, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... GPC3(2719)

General description

Glypican 3 (GPC3), whose gene is associated with Simpson-Golabi-Behmel syndrome, is a heparin sulfate proteoglycan that is anchored to cell-surfaces via glycosylphosphatidylinositol (GPI). Overexpression of glypican 3 occurs in hepatocellular carcinomas (HCC). The silencing of glypican 3 expression leads to apoptosis in HCC.

Specificity

Anti-GPC3 polyclonal antibody reacts with canine, bovine, human, mouse, and rat glypican 3 proteins.

Immunogen

The immunogen for anti-GPC3 antibody: synthetic peptide derected towards the middle region of human GPC3

Application

Anti-GPC3 polyclonal antibody is used to tag glypican 3 protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe in cancer diagnosis to detect the presence of glypican 3 in hepatocellular carcinomas (HCC).

Sequence

Synthetic peptide located within the following region: FSTIHDSIQYVQKNAGKLTTTIGKLCAHSQQRQYRSAYYPEDLFIDKKVL

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service