Skip to Content
Merck
All Photos(1)

Key Documents

AV37377

Sigma-Aldrich

Anti-ISL2 antibody produced in rabbit

IgG fraction of antiserum

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€287.00

€287.00


Estimated to ship on27 April 2025



Select a Size

Change View
100 μL
€287.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€287.00


Estimated to ship on27 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... ISL2(104360)

General description

Insulin gene enhancer protein, ISL-2, is a LIM homeobox domain transcription factor. Human ISL-2 is a 39 kDa protein found expressed in the nucleus of numerous cell types including motorneurons early in development of the spinal column.

Immunogen

Synthetic peptide directed towards the C terminal region of mouse ISL2

Biochem/physiol Actions

Isl2 specifies RGC laterality by repressing an ipsilateral pathfinding program unique to VTC RGCs and involving Zic2 and EphB1. This genetic hierarchy controls binocular vision.

Sequence

Synthetic peptide located within the following region: VIRVWFQNKRCKDKKKSILMKQLQQQQHSDKASFQGLTGTPLVAGSPIGH

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua P Thaler et al.
Neuron, 41(3), 337-350 (2004-02-10)
LIM homeobox genes have a prominent role in the regulation of neuronal subtype identity and distinguish motor neuron subclasses in the embryonic spinal cord. We have investigated the role of Isl-class LIM homeodomain proteins in motor neuron diversification using mouse

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service