Skip to Content
Merck
All Photos(1)

Key Documents

AV35650

Sigma-Aldrich

Anti-SLC18A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Solute carrier family 18 (vesicular monoamine), member 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€374.00

€374.00


Estimated to ship on21 May 2025



Select a Size

Change View
100 μL
€374.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€374.00


Estimated to ship on21 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC18A1(6570)

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC18A1

Biochem/physiol Actions

Solute carrier family 18 (vesicular monoamine), member 1 (SLC18A1) is a vesicular monamine transporter that is involved in the transport of monoamine neurotransmitters. It accumulates cytosolic monoamines into vesicles based on the proton gradient. Variations in SLC18A1 transporter have been observed in neuropsychiatric disorders.

Sequence

Synthetic peptide located within the following region: MNDTASTIPPPATEAISAHKNNCLQGTGFLEEEITRVGVLFASKAVMQLL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed M Khalifa et al.
Neuro endocrinology letters, 33(5), 546-551 (2012-10-24)
Although single nucleotide polymorphisms of the human vesicular monoamine transporter 1 (hVMAT1) gene SLC18A1 have been associated with neuropsychiatric disorders, there is limited information on the function of naturally occurring hVMAT1 variant proteins. This study evaluated transport activity of full
Falk W Lohoff et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 31(12), 2739-2747 (2006-08-29)
The vesicular monoamine transporter 1 gene (VMAT1/SLC18A1) maps to the shared bipolar disorder (BPD)/schizophrenia (SZ) susceptibility locus on chromosome 8p21. Vesicular monoamine transporters are involved in transport of monoamine neurotransmitters which have been postulated to play a relevant role in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service