Skip to Content
Merck
All Photos(2)

Key Documents

AV35259

Sigma-Aldrich

Anti-P2RX2 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Purinergic receptor P2X, ligand-gated ion channel, 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€287.00

€287.00


Estimated to ship on31 May 2025



Select a Size

Change View
100 μL
€287.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€287.00


Estimated to ship on31 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... P2RX2(22953)

General description

P2RX2 (P2X2) is a purinoceptor that functions as an ATP-gated ion channel. The ectodomain movements of this receptor have been analyzed using disulphide locking. In propyl-methanethiosulfonate-attached P2X2 receptors; lipid intercalation can be facilitated between the transmembrane domains during channel opening.
Rabbit Anti-P2RX2 antibody recognizes human and bovine P2RX2.

Immunogen

Synthetic peptide directed towards the middle region of human P2RX2

Application

Rabbit Anti-P2RX2 is suitable for western blot (0.6 μg/ml) and IHC (4-8 μg/ml) applications.

Biochem/physiol Actions

P2RX2 belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel. Binding to ATP mediates synaptic transmission between neurons and from neurons to smooth muscle. Six transcript variants encoding six distinct isoforms have been identified for this gene.

Sequence

Synthetic peptide located within the following region: LIKNSIHYPKFHFSKGNIADRTDGYLKRCTFHEASDLYCPIFKLGFIVEK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga Stelmashenko et al.
The Journal of biological chemistry, 289(14), 9909-9917 (2014-02-12)
The ectodomain of the P2X receptor is formed mainly from two- or three-stranded β-sheets provided symmetrically by each of the three subunits. These enclose a central cavity that is closed off furthest from the plasma membrane (the turret) and that
Simon W Rothwell et al.
The Journal of biological chemistry, 289(2), 618-626 (2013-11-26)
The ionic pore of the P2X receptor passes through the central axis of six transmembrane (TM) helices, two from each of three subunits. Val(48) and Ile(328) are at the outer end of TM1 and TM2, respectively. Homology models of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service