Skip to Content
Merck
All Photos(1)

Key Documents

AV34379

Sigma-Aldrich

Anti-ELK4 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-ELK4, ETS-domain protein (SRF accessory protein 1)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€326.00

€326.00


Estimated to ship on29 May 2025



Select a Size

Change View
100 μL
€326.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€326.00


Estimated to ship on29 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

47 kDa

species reactivity

mouse, horse, bovine, human, pig, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ELK4(2005)

General description

ETS domain-containing protein Elk-4 is a member of the Ets family of transcription factors. Elk-4 was recently shown to be a target for the androgen receptor in prostate cancer cells.

Immunogen

Synthetic peptide directed towards the C terminal region of human ELK4

Biochem/physiol Actions

The ELK4 gene is a member of the Ets family of transcription factors and of the ternary complex factor (TCF) subfamily. Proteins of the TCF subfamily form a ternary complex by binding to the the serum response factor and the serum reponse element in the promoter of the c-fos proto-oncogene. The protein encoded by ELK4 is phosphorylated by the kinases, MAPK1 and MAPK8.

Sequence

Synthetic peptide located within the following region: PVAPLSPARLQGANTLFQFPSVLNSHGPFTLSGLDGPSTPGPFSPDLQKT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Makkonen et al.
Oncogene, 27(36), 4865-4876 (2008-05-13)
Transcriptional control by androgens via androgen receptor (AR) is strongly involved in prostate cancer development, but the critical target genes have remained elusive. We have characterized E twenty-six-like transcription factor 4 (ELK4) (also known as serum response factor accessory protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service