Skip to Content
Merck
All Photos(1)

Documents

AV100888

Sigma-Aldrich

Anti-TRIM22 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-GPSTAF50, Anti-RNF94, Anti-STAF50, Anti-Tripartite motif-containing 22

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

57 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIM22(10346)

Immunogen

Synthetic peptide directed towards the N terminal region of human TRIM22

Biochem/physiol Actions

TRIM22 exerts anti-viral activity against RNA viruses and is stimulated by interferon-α. It restricts the replication of influenza A virus by interacting with viral nucleoprotein followed by ubiquitination and proteasome-dependent degradation. TRIM22 also restricts the replication of encephalomyocarditis virus (EMCV), hepatitis B virus (HBV), and human immunodeficiency virus type 1 (HIV-1). It has been implicated in cellular differentiation and proliferation of certain cancers and autoimmune reactions.

Sequence

Synthetic peptide located within the following region: LANIVERVKEVKMSPQEGQKRDVCEHHGKKLQIFCKEDGKVICWVCELSQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clayton J Hattlmann et al.
Molecular biology international, 2012, 153415-153415 (2012-06-01)
The tripartite motif (TRIM) family of proteins is an evolutionarily ancient group of proteins with homologues identified in both invertebrate and vertebrate species. Human TRIM22 is one such protein that has a dynamic evolutionary history that includes gene expansion, gene
Andrea Di Pietro et al.
Journal of virology, 87(8), 4523-4533 (2013-02-15)
Tripartite motif (TRIM) protein superfamily members are emerging as important effectors of the innate immune response against viral infections. In particular, TRIM22 was reported to exert antiviral activity against RNA viruses, such as hepatitis B virus (HBV), encephalomyocarditis virus (ECMV)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service