Skip to Content
Merck
All Photos(4)

Documents

AV100816

Sigma-Aldrich

Anti-NR2F2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ARP1, Anti-COUP-TFII, Anti-COUPTFB, Anti-MGC117452, Anti-Nuclear receptor subfamily 2, group F, member 2, Anti-SVP40, Anti-TFCOUP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

45 kDa

species reactivity

dog, pig, human, mouse, rabbit, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR2F2(7026)

Immunogen

Synthetic peptide directed towards the N terminal region of human NR2F2

Application

Anti-NR2F2 antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml.

Biochem/physiol Actions

NR2F2 is an orphan nuclear receptor that plays an important role in metabolism and development. The crosstalk between NR2F2 and the transcription factor HNF4α is involved in regulation of insulin secretion and maintenance of glucose homeostasis. NR2F2 collaborates with OCT4 and miR-302 in differentiation of human embryonic stem cells and specification of neural ectoderm during development.

Sequence

Synthetic peptide located within the following region: KVGMRREAVQRGRMPPTQPTHGQFALTNGDPLNCHSYLSGYISLLLRAEP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Boutant et al.
PloS one, 7(5), e35810-e35810 (2012-05-19)
The Nuclear Receptor 2F2 (NR2F2/COUP-TFII) heterozygous knockout mice display low basal insulinemia and enhanced insulin sensitivity. We previously established that insulin represses NR2F2 gene expression in pancreatic β-cells. The cis-regulatory region of the NR2F2 promoter is unknown and its influence
Alessandro Rosa et al.
The EMBO journal, 30(2), 237-248 (2010-12-15)
Multiple levels of control are in play to regulate pluripotency and differentiation in human embryonic stem cells (hESCs). At the transcriptional level, the core factors OCT4, NANOG and SOX2 form a positive autoregulatory loop that is pivotal for maintaining the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service