This enzyme phosphorylates and inactivates protein phosphatase 2A. The funtion appears to be related to the catalytic domain of p21-activated p65(PAK) protein kinase which is produced in apoptotic cells. AK is also involved in cytoskeleton organization and other signal transduction processes.
Unit Definition
One unit is the amount of enzyme that will incorporate 1 nmole of phosphoryl groups using myelin basic protein as a substrate per minute at pH 7 at 30 deg C.
Physical form
50 ng per vial in 10 μl of 50 mM Tris-HCl, pH 7.0, containing 14 mM β-Mercaptoethanol, 1 mM benzamidine, 0.1 mM PMSF, 1 mM EDTA, 65% ethyleneglycol, and 0.1% Brij® 35.
Legal Information
Brij is a registered trademark of Croda International PLC
Frontiers in immunology, 13, 959495-959495 (2022-08-16)
Macrophages are involved in hepatocyte steatosis and necroinflammation and play an important role in the pathogenesis of non-alcoholic fatty liver disease (NAFLD). Impaired autophagy function (decreased autophagy or blocked autophagic flow) leads to cell damage and death and promotes NAFLD
The Journal of biological chemistry, 296, 100667-100667 (2021-04-18)
The epoxyeicosatrienoic acid (EET) exerts beneficial effects on insulin resistance and/or hypertension. EETs could be readily converted to less biological active diols by soluble epoxide hydrolase (sEH). However, whether sEH inhibition can ameliorate the comorbidities of insulin resistance and hypertension
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.