Skip to Content
Merck
All Photos(1)

Key Documents

A7661

Sigma-Aldrich

N-Acetyl-D-glucosamine 6-sulfate sodium salt

≥97% (HPAE)

Synonym(s):

GlcNAc-6S

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H14NO9SNa
CAS Number:
Molecular Weight:
323.25
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

description

Mixed anomers

Assay

≥97% (HPAE)

form

powder

storage temp.

2-8°C

SMILES string

[Na+].CC(=O)N[C@H]1C(O)O[C@H](COS([O-])(=O)=O)[C@@H](O)[C@@H]1O

InChI

1S/C8H15NO9S.Na/c1-3(10)9-5-7(12)6(11)4(18-8(5)13)2-17-19(14,15)16;/h4-8,11-13H,2H2,1H3,(H,9,10)(H,14,15,16);/q;+1/p-1/t4-,5-,6-,7-,8?;/m1./s1

InChI key

CBUJZKTVEFVBBG-FROKLYQUSA-M

Looking for similar products? Visit Product Comparison Guide

Application

N-Acetyl-D-glucosamine 6-sulfate (GlcNAc-6S) is used as a substrate and inhibitor to identify, differentiate and characterize various sulfatase(s), especially N-acetylglucosamine-6-sulphatase(s).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew J Nok et al.
Journal of biochemical and molecular toxicology, 17(1), 59-66 (2003-03-05)
The venom of Naja nigricolis was found to contain a high level of the enzyme aryl sulfatase. The enzyme was isolated from the venom of N. nigriclois and purified to electrophoretic homogeneity by gel chromatography on Sephadex G-100, DEAE-cellulose, and
Shengyuan Xu et al.
The Biochemical journal, 387(Pt 3), 841-847 (2004-12-15)
A 75 kDa protein was purified to homogeneity from granule extracts of normal human granulocytes using Sephadex G-75 chromatography, Mono-S cation exchange chromatography and chromatofocusing. The protein consisted of one chain with a molecular mass of 75 kDa, as determined
Andrea Mok et al.
Genomics, 81(1), 1-5 (2003-02-08)
Mucopolysaccharidosis type IIID (MPS IIID; Sanfilippo syndrome type D; MIM 252940) is caused by deficiency of the activity of N-acetylglucosamine-6-sulfatase (GNS), which is normally required for degradation of heparan sulfate. The clinical features of MPS IIID include progressive neurodegeneration, with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service