Skip to Content
Merck
All Photos(1)

Documents

A7475

Sigma-Aldrich

Anthopleurin-A trifluoroacetate salt

>88% (HPLC)

Synonym(s):

AP-A trifluoroacetate salt, Anthopleura toxin A trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C220H326N64O67S6 · xC2HF3O2
Molecular Weight:
5131.72 (free base basis)
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

>88% (HPLC)

form

solid

storage temp.

−20°C

Amino Acid Sequence

Gly-Val-Ser-Cys-Leu-Cys-Asp-Ser-Asp-Gly-Pro-Ser-Val-Arg-Gly-Asn-Thr-Leu-Ser-Gly-Thr-Leu-Trp-Leu-Tyr-Pro-Ser-Gly-Cys-Pro-Ser-Gly-Trp-His-Asn-Cys-Lys-Ala-His-Gly-Pro-Thr-Ile-Gly-Trp-Cys-Cys-Lys-Gln [Disulfide bridges: 4-46, 6-36, 29-47]

General description

Synthetic peptide toxin that was originally isolated from the sea anemone, Anthopleura xantho­grammica.

Application

Anthopleurin-A is a toxin used to study the gating mechanisms of sodium channels. Anthopleurin-A slows the repolarization phase of nerve and muscle action potentials by inactivating the sodium channel.

Biochem/physiol Actions

Anthopleurin-A slows the repolarization phase of nerve and muscle action potentials by inactivating the sodium channel. Anthopleurin-A shows a preference towards cardiac channels over the neuronal sodium channels. It is a toxin used to study the gating mechanisms of sodium channels.
Shown to have inotropic effects and not chronotropic effects on mammalian heart preparations.

Other Notes

supplied as trifluoroacetate salt

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P K Pallaghy et al.
Biochemistry, 34(11), 3782-3794 (1995-03-21)
The three-dimensional structure in aqueous solution of the 49-residue polypeptide anthopleurin-A (AP-A), from the sea anemone Anthopleura xanthogrammica, has been determined from 1H NMR data. A restraint set consisting of 411 interproton distance restraints inferred from NOEs and 19 backbone
Y Pichon
Journal of physiology, Paris, 89(4-6), 171-180 (1995-01-01)
The physiological function of the axon is to conduct short all-or-none action potentials from their site of initiation (usually the cell body) to the synapse. To ensure this function, both passive and active biophysical properties of the axons are tuned
M J Scanlon et al.
Protein science : a publication of the Protein Society, 3(7), 1121-1124 (1994-07-01)
Anthopleurin-A (AP-A) is a member of a family of sea anemone-derived polypeptides that interact with sodium channels in a voltage-dependent manner, producing a positive inotropic effect on the mammalian heart. There has been considerable interest in this molecule as a
W Shimizu et al.
Circulation, 96(6), 2038-2047 (1997-10-10)
This study examines the contribution of transmural heterogeneity of transmembrane activity to phenotypic T-wave patterns and the effects of pacing and of sodium channel block under conditions mimicking HERG and SCN5A defects linked to the congenital long-QT syndrome (LQTS). A
S G Priori et al.
Pacing and clinical electrophysiology : PACE, 20(8 Pt 2), 2052-2057 (1997-08-01)
The long QT syndrome (LQTS) is a familial disease characterized by prolonged ventricular repolarization and high incidence of malignant ventricular tachyarrhythmias often occurring in conditions of adrenergic activation. Recently, the genes for the LQTS inked to chromosomes 3 (LQT3), 7

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service