Skip to Content
Merck
All Photos(1)

Key Documents

A6882

Sigma-Aldrich

Atractyloside potassium salt

≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H44K2O16S2
CAS Number:
Molecular Weight:
802.99
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

solubility

H2O: 20 mg/mL

SMILES string

[K+].[K+].[H][C@@]12CC[C@@]3([H])[C@]4(C)C[C@@H](C[C@@H](C(O)=O)[C@@]4([H])CC[C@]3(C1)[C@@H](O)C2=C)O[C@@H]5O[C@H](CO)[C@@H](OS([O-])(=O)=O)[C@H](OS([O-])(=O)=O)[C@H]5OC(=O)CC(C)C

Biochem/physiol Actions

An extremely toxic glycoside isolated from thistles. Inhibits oxidative phosphorylation by blocking the transfer of adenosine nucleotides through the mitochondrial membrane.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian A Barrientos et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(3), 966-978 (2011-01-21)
Axonal degeneration is an active process that has been associated with neurodegenerative conditions triggered by mechanical, metabolic, infectious, toxic, hereditary and inflammatory stimuli. This degenerative process can cause permanent loss of function, so it represents a focus for neuroprotective strategies.
Rashel Mizrahi et al.
Biochimica et biophysica acta, 1840(12), 3404-3412 (2014-09-16)
Mitochondria are the major source of ATP to power sperm motility. Phosphorylation of mitochondrial proteins has been proposed as a major regulatory mechanism for mitochondrial bioenergetics. Sperm motility was measured by a computer-assisted analyzer, protein detection by western blotting, membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service