All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H13NO2 · C7H8O3S
CAS Number:
Molecular Weight:
351.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
Assay
≥98% (TLC)
form
powder
color
white
application(s)
peptide synthesis
storage temp.
−20°C
SMILES string
Cc1ccc(cc1)S(O)(=O)=O.NCCC(=O)OCc2ccccc2
InChI
1S/C10H13NO2.C7H8O3S/c11-7-6-10(12)13-8-9-4-2-1-3-5-9;1-6-2-4-7(5-3-6)11(8,9)10/h1-5H,6-8,11H2;2-5H,1H3,(H,8,9,10)
InChI key
FRHWYVGCFUQMJR-UHFFFAOYSA-N
Biochem/physiol Actions
β-Alanine benzyl ester (β-Ala-OBzl) is used in the biochemical synthesis of β-alanyl peptides via aminopeptidase(s).
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jiro Arima et al.
Applied and environmental microbiology, 72(6), 4225-4231 (2006-06-06)
Dipeptide synthesis by aminopeptidase from Streptomyces septatus TH-2 (SSAP) was demonstrated using free amino acid as an acyl donor and aminoacyl methyl ester as an acyl acceptor in 98% methanol (MeOH). SSAP retained its activity after more than 100 h
Jiro Arima et al.
Journal of biotechnology, 147(1), 52-58 (2010-03-23)
Synthesis of beta-alanine (beta-Ala) containing dipeptide using S9 aminopeptidase from Streptomyces thermocyaneoviolaceus NBRC14271 (S9AP-St) was demonstrated with beta-Ala-benzyl ester (-OBzl) and various L-aminoacyl derivatives. For synthesis of beta-Ala-containing dipeptide, beta-Ala-OBzl was used preferentially as the acyl donor for S9AP-St, producing
Linda Clijsters et al.
Molecular cell, 74(6), 1264-1277 (2019-05-28)
E2F1, E2F2, and E2F3A, the three activators of the E2F family of transcription factors, are key regulators of the G1/S transition, promoting transcription of hundreds of genes critical for cell-cycle progression. We found that during late S and in G2
Lin Deng et al.
Molecular cell, 73(5), 915-929 (2019-03-09)
DNA replication errors generate complex chromosomal rearrangements and thereby contribute to tumorigenesis and other human diseases. One mechanism that triggers these errors is mitotic entry before the completion of DNA replication. To address how mitosis might affect DNA replication, we
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service