All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H19N3O4
CAS Number:
Molecular Weight:
293.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
technique(s)
ligand binding assay: suitable
storage temp.
−20°C
SMILES string
CC(C)C[C@H](NC(C)=O)C(=O)Nc1ccc(cc1)N(=O)=O
InChI
1S/C14H19N3O4/c1-9(2)8-13(15-10(3)18)14(19)16-11-4-6-12(7-5-11)17(20)21/h4-7,9,13H,8H2,1-3H3,(H,15,18)(H,16,19)/t13-/m0/s1
InChI key
SLJKQHDBYBGOMH-ZDUSSCGKSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Federica Parravicini et al.
PloS one, 8(2), e56254-e56254 (2013-03-05)
Acyl aminoacyl peptidases are two-domain proteins composed by a C-terminal catalytic α/β-hydrolase domain and by an N-terminal β-propeller domain connected through a structural element that is at the N-terminus in sequence but participates in the 3D structure of the C-domain.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service