Skip to Content
Merck
All Photos(1)

Key Documents

96647

Sigma-Aldrich

Firoin from Rhodothermus marinus

~80% (TLC)

Synonym(s):

(2R)-O2-(β-D-mannopyranosyl)glyceric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O9
CAS Number:
Molecular Weight:
268.22
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

~80% (TLC)

form

crystals

impurities

~10% potassium chloride

SMILES string

OC[C@H]1O[C@@H](O[C@H](CO)C(O)=O)[C@@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C9H16O9/c10-1-3-5(12)6(13)7(14)9(17-3)18-4(2-11)8(15)16/h3-7,9-14H,1-2H2,(H,15,16)/t3-,4-,5-,6+,7+,9+/m1/s1

InChI key

DDXCFDOPXBPUJC-MTXRGOKVSA-N

Application

Firoin from Rhodothermus marinus, an extremolyte, may be used in the development of potential new formulations to enhance the stability of retroviral and adenoviral vectors.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Georg Lentzen et al.
Applied microbiology and biotechnology, 72(4), 623-634 (2006-09-08)
Extremophilic microorganisms have adopted a variety of ingenious strategies for survival under high or low temperature, extreme pressure, and drastic salt concentrations. A novel application area for extremophiles is the use of "extremolytes," organic osmolytes from extremophilic microorganisms, to protect
Pedro E Cruz et al.
Biotechnology progress, 22(2), 568-576 (2006-04-08)
In the past decade there has been an increase in the application of viral vectors in the laboratory and clinical trials of human gene therapy, retroviral and adenoviral vectors among the most used. However, the limited stability of these vectors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service