Skip to Content
Merck
All Photos(1)

Key Documents

93287

Sigma-Aldrich

Trizma® hydrochloride

BioUltra, for luminescence, ≥99.0% (AT)

Synonym(s):

TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Beilstein:
3675235
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

grade

for luminescence

product line

BioUltra

Assay

≥99.0% (AT)

form

powder or crystals

impurities

insoluble matter, passes filter test

ign. residue

≤0.2% (as SO4)

loss

≤0.2% loss on drying, 110 °C

pH

2.5-4.0 (25 °C, 4 M in H2O)

useful pH range

7.0-9.0

pKa (25 °C)

8.1

mp

150-152 °C

solubility

H2O: 4 M at 20 °C, clear, colorless

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

fluorescence

λex 200-600 nm; λem 300-800 nm (corresponds)

λ

4 M in H2O

UV absorption

λ: 260 nm Amax: 0.12
λ: 280 nm Amax: 0.08

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Easily compare specifications for Trizma HCl products with the Trizma HCl specification table.
The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution. For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tibor Maliar et al.
Chemistry & biodiversity, 12(11), 1678-1695 (2015-11-17)
The content of biogenic compounds and the biological activities of barley (Hordeum vulgare L.)-grain extracts was evaluated. The sufficiently large and heterogeneous set of barley genotypes (100 accessions) enabled the selection of special genotypes interesting for potential industrial, pharmaceutical, and
Yingning Zheng et al.
Analytica chimica acta, 834, 45-50 (2014-06-15)
Our present work aimed at developing a pseudo triple-enzyme cascade electrocatalytic electrochemical aptasensor for determination of thrombin with the amplification of alcohol dehydrogenase (ADH)-Pt-Pd nanowires bionanocomposite and hemin/G-quadruplex structure that simultaneously acted as NADH oxidase and HRP-mimicking DNAzyme. With the
Michiya Matsusaki et al.
Journal of biomedical materials research. Part A, 103(10), 3386-3396 (2015-04-09)
We developed a human skin equivalent (HSE) containing blood and lymph-like capillary networks using a cell coating technique, which is a rapid fabrication technology of three-dimensional cellular constructs by cell surface coating using layer-by-layer assembled nanofilms of extracellular matrices. The
Illana Gozes et al.
Journal of Alzheimer's disease : JAD, 40 Suppl 1, S23-S36 (2014-02-08)
NAP (NAPVSIPQ, davunetide) is a microtubule stabilizing peptide drug candidate. Here, we set out to identify NAP-like peptides that provide neuroprotection and reduce tau pathology. NAP-like peptides were derived using publically available search engines, which identified sequence homologies in the
Shao-Hua Wu et al.
Biosensors & bioelectronics, 73, 41-46 (2015-06-05)
A new gold particles modified heated copper disk electrode (Au-HCuDE) with direct current was fabricated. The hemin/G-quadruplex horseradish peroxidase-mimicking DNAzyme (HRP-DNAzyme) was self-assembled on the heated electrode and resulted in a new biosensor denoted as HRP-DNAzyme/Au-HCuDE. By controlling the temperature

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service