Skip to Content
Merck
All Photos(1)

Key Documents

91924

Sigma-Aldrich

Tentagel M OH Microspheres

extent of labeling: ~0.2 mmol/g loading

Synonym(s):

O-(2-Hydroxyethyl)polyethylene glycol grafted to polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

form

powder

extent of labeling

~0.2 mmol/g loading

particle size

10 μm

storage temp.

2-8°C

Application

Tentagel beads are functionalized polystyrene-polyethylene glycol (PS-PEG) beads that may be used in the development of combinatorial libraries for high throughput analysis.

Legal Information

TentaGel is a trademark of Rapp Polymere GmbH

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gary L Juskowiak et al.
Journal of combinatorial chemistry, 10(5), 726-731 (2008-07-29)
Cysteine-rich peptides are valued as tags for biarsenical fluorophores and as environmentally important reagents for binding toxic heavy metals. Due to the inherent difficulties created by cysteine, the power of one-bead one-compound (OBOC) libraries has never been applied to the
H Weinberger et al.
Archiv der Pharmazie, 330(4), 109-111 (1997-04-01)
Peptides from small combinatorial libraries, covalently attached to polymeric TentaGel beads, can be directly sequenced using amino acid analysis. For libraries with restricted diversity, generated by the split-mix synthesis method, the amino acids on a selected single bead identified by
Hyoung Gee Baek et al.
Journal of combinatorial chemistry, 11(1), 91-102 (2008-12-09)
The one-bead one-compound (OBOC) combinatorial library method enables the rapid generation and screening of millions of discrete chemical compounds on beads. Most of the OBOC screening methods require the library compounds to remain tethered to the bead during screening process.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service