Skip to Content
Merck
All Photos(1)

Key Documents

83636

Sigma-Aldrich

Resorufin acetate

suitable for fluorescence, ≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9NO4
CAS Number:
Molecular Weight:
255.23
Beilstein:
246139
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98.0% (TLC)

form

powder or crystals

mp

223 °C (lit.)

solubility

DMF: soluble
DMSO: soluble
alcohols: soluble

fluorescence

λex 500 nm; λem 593 nm in 0.1 M phosphate pH 8.0 (chymotrypsin)
λex 540 nm; λem 590 nm in DMSO
λex 571 nm; λem 584 nm (Reaction product)

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccc2N=C3C=CC(=O)C=C3Oc2c1

InChI

1S/C14H9NO4/c1-8(16)18-10-3-5-12-14(7-10)19-13-6-9(17)2-4-11(13)15-12/h2-7H,1H3

InChI key

UJWKHDKBOVPINX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

suitable as fluorogenic substrate for hydrolytic enzymes (cellulases, chymotrypsin)

Other Notes

Studies on the esterase activity of cytosolic aldehyde dehydrogenase

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Myung Gil Choi et al.
Organic letters, 12(7), 1468-1471 (2010-03-13)
The acetate derivatives of fluorescein and resorufin exhibited a prominent turn-on type signaling behavior toward BO(3)(-) ions over other common anions. Signaling is based on the selective deprotection of acetate groups by perborate, which resulted in significant chromogenic and fluorogenic
The action of cytosolic aldehyde dehydrogenase on resorufin acetate.
T M Kitson et al.
Advances in experimental medicine and biology, 414, 201-208 (1997-01-01)
T M Kitson
Biochimica et biophysica acta, 1385(1), 43-52 (1998-06-19)
Resorufin bromoacetate is a substrate that is rapidly hydrolysed by chymotrypsin. The reaction shows a pre-steady-state burst phase that may be observed by stopped flow spectrophotometry if precautions are taken against spontaneous hydrolysis of the substrate. The strongly activating effect
T M Kitson et al.
The Biochemical journal, 322 ( Pt 3), 701-708 (1997-03-15)
Resorufin acetate is a very good substrate for sheep liver cytosolic aldehyde dehydrogenase, both from the point of view of practical spectrophotometry and in terms of information provided about the nature of the catalysis shown by this enzyme. p-Nitrophenyl (PNP)
Mohit P Mathew et al.
Chembiochem : a European journal of chemical biology, 18(13), 1204-1215 (2017-02-22)
This report describes the metabolic glycoengineering (MGE) of intracellular esterase activity in human colon cancer (LS174T) and Chinese hamster ovary (CHO) cells. In silico analysis of carboxylesterases CES1 and CES2 suggested that these enzymes are modified with sialylated N-glycans, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service