Skip to Content
Merck
All Photos(1)

Key Documents

83132

Sigma-Aldrich

Pyrogallol

purum, meets analytical specification of Ph. Helv. 7

Synonym(s):

Pyrogallolum, 1,2,3-Trihydroxybenzene, 2,3-Dihydroxyphenol, Pyrogallic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(OH)3
CAS Number:
Molecular Weight:
126.11
Beilstein:
907431
EC Number:
MDL number:
UNSPSC Code:
12352200

vapor density

4.4 (vs air)

vapor pressure

10 mmHg ( 167.7 °C)

grade

purum

quality

meets analytical specification of Ph. Helv. 7

bp

309 °C (lit.)

mp

133-134 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

Oc1cccc(O)c1O

InChI

1S/C6H6O3/c7-4-2-1-3-5(8)6(4)9/h1-3,7-9H

InChI key

WQGWDDDVZFFDIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drew A Fowler et al.
Chemical communications (Cambridge, England), 49(18), 1802-1804 (2013-01-30)
The crystal structure of a cocrystal comprising a complex of a bis(imidazolium) ionic liquid hosted by pyrogallol[4]arene (PgC) macrocycles is reported. Introduction of the dicationic species yields complexation by two PgC macrocycles, the resulting bilayer structure formed from alternately arranged
Adelaide U P Hain et al.
Journal of structural biology, 180(3), 551-562 (2012-09-18)
The autophagy-related proteins are thought to serve multiple functions in Plasmodium and are considered essential to parasite survival and development. We have studied two key interacting proteins, Atg8 and Atg3, of the autophagy pathway in Plasmodium falciparum. These proteins are
National Toxicology Program technical report series, (574)(574), 1-167 (2013-03-23)
The current main commercial use of pyrogallol is the production of pharmaceuticals and pesticides. In analytical chemistry, pyrogallol is used as a complexing agent, reducing agent, and, in alkaline solution, as an indicator of gaseous oxygen. Pyrogallol was nominated for
Suzanne J P L van den Berg et al.
Chemical research in toxicology, 25(11), 2352-2367 (2012-09-21)
The present study describes physiologically based kinetic (PBK) models for the alkenylbenzene elemicin (3,4,5-trimethoxyallylbenzene) in rat and human, based on the PBK models previously developed for the structurally related alkenylbenzenes estragole, methyleugenol, and safrole. Using the newly developed models, the
Shaolin Mu et al.
The journal of physical chemistry. B, 116(41), 12567-12573 (2012-09-27)
Electrochemical oxidation of pyrogallol in a pH 5.0 phosphate buffer was carried out on a reduced graphene oxide/glassy carbon (RGO/GC) electrode. Reduced graphene oxide plays an important role in catalyzing the electrochemical oxidation of pyrogallol. A deep yellow film deposited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service