Skip to Content
Merck
All Photos(1)

Key Documents

51292

Sigma-Aldrich

Hemoglobin from bovine blood

≥85% (HPCE)

Synonym(s):

Bovine hemoglobin, Hb, Methemoglobin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42030116

biological source

bovine blood

Assay

≥85% (HPCE)
~95% protein basis

mol wt

Mr ~64500

loss

≤5% loss on drying

UniProt accession no.

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

The solubility of α-elastin has been applied to construction of elastin-mimetic biomaterials.

Biochem/physiol Actions

Oxygen transporter, NO scavenger
Oxygen transporter. The Fe2+/Fe3+ balance is a physiological indicator of blood oxygenation; deoxygenated hemoglobin accessorizes a feedback loop by reducing nitrite to NO, a vasodilator which enhances blood flow to oxygen-deprived tissues.

Caution

Since native hemoglobin is readily oxidized in air, these preparations may be predominantly methemoglobin.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masayo Sawa et al.
Neuropsychobiology, 67(2), 103-110 (2013-02-15)
Only a few studies have examined the relationships between affective symptoms, cognitive function (e.g. verbal fluency), quality of life (QOL), and brain activation in a nonclinical population. The aim of the present study was to assess these relationships and examine
Toru Yamada et al.
PloS one, 7(11), e50271-e50271 (2012-11-28)
In conventional functional near-infrared spectroscopy (fNIRS), systemic physiological fluctuations evoked by a body's motion and psychophysiological changes often contaminate fNIRS signals. We propose a novel method for separating functional and systemic signals based on their hemodynamic differences. Considering their physiological
N F Schreiter et al.
RoFo : Fortschritte auf dem Gebiete der Rontgenstrahlen und der Nuklearmedizin, 185(4), 358-366 (2013-03-16)
To investigate whether changes in hemodynamics induced by Valsalva maneuver can be exploited for detecting and characterizing breast lesions by optical mammography. 30 women underwent optical imaging of the breast using a DYNOT 232 system and performing Valsalva maneuvers prior
R Bokiniec et al.
Advances in medical sciences, 57(2), 348-355 (2012-11-20)
The aim of this study was to determine brain oxygenation in full-term and preterm neonates using near infrared spectroscopy. A total of 88 full-term and preterm newborn infants without hypoxic-ischaemic disorders admitted to the NICU were examined using NIRS on
Jae G Kim et al.
Journal of biomedical optics, 17(10), 105005-105005 (2012-12-12)
Noninvasive near infrared spectroscopy measurements were performed to monitor cyanide (CN) poisoning and recovery in the brain region and in foreleg muscle simultaneously, and the effects of a novel CN antidote, sulfanegen sodium, on tissue hemoglobin oxygenation changes were compared

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service